missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-4,5-dichloro-1H-imidazole, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
Specifications
Specifications
| CAS Min % | 97.0 |
| Assay Percent Range | 97% |
| Packaging | Amber Glass Bottle |
| Quantity | 1 g |
| Formula Weight | 215.86 |
| Percent Purity | 97% |
| Chemical Name or Material | 2-Bromo-4, 5-dichloro-1H-imidazole |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction