Learn More
2-Bromo-4,6-dichloroaniline, 98+%, Thermo Scientific Chemicals
£61.90 - £171.00
Chemical Identifiers
CAS | 697-86-9 |
---|---|
Molecular Formula | C6H4BrCl2N |
Molecular Weight (g/mol) | 240.909 |
MDL Number | MFCD00040936 |
InChI Key | DTPADCOGQUOGHT-UHFFFAOYSA-N |
Synonym | pubchem3605, acmc-209ob3, 6-bromo-2,4-dichloroaniline, 2,4-dichloro-6-bromoaniline, 2-bromo-4,6-dichloro-aniline, 6-bromo-2,4-dichlorophenylamine, benzenamine,2-bromo-4,6-dichloro, 2-bromo-4,6-dichloroaniline, 2-bromanyl-4,6-bis chloranyl aniline |
PubChem CID | 2756901 |
IUPAC Name | 2-bromo-4,6-dichloroaniline |
SMILES | C1=C(C=C(C(=C1Cl)N)Br)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11468357
|
Thermo Scientific Alfa Aesar
A18436.14 |
25 g |
£61.90
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11478357
|
Thermo Scientific Alfa Aesar
A18436.22 |
100 g |
£171.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
697-86-9 | |
240.909 | |
DTPADCOGQUOGHT-UHFFFAOYSA-N | |
2756901 | |
C1=C(C=C(C(=C1Cl)N)Br)Cl |
C6H4BrCl2N | |
MFCD00040936 | |
pubchem3605, acmc-209ob3, 6-bromo-2,4-dichloroaniline, 2,4-dichloro-6-bromoaniline, 2-bromo-4,6-dichloro-aniline, 6-bromo-2,4-dichlorophenylamine, benzenamine,2-bromo-4,6-dichloro, 2-bromo-4,6-dichloroaniline, 2-bromanyl-4,6-bis chloranyl aniline | |
2-bromo-4,6-dichloroaniline |
Specifications
697-86-9 | |
C6H4BrCl2N | |
2089686 | |
DTPADCOGQUOGHT-UHFFFAOYSA-N | |
2-bromo-4,6-dichloroaniline | |
2756901 | |
≥98% |
78°C to 82°C | |
MFCD00040936 | |
pubchem3605, acmc-209ob3, 6-bromo-2,4-dichloroaniline, 2,4-dichloro-6-bromoaniline, 2-bromo-4,6-dichloro-aniline, 6-bromo-2,4-dichlorophenylamine, benzenamine,2-bromo-4,6-dichloro, 2-bromo-4,6-dichloroaniline, 2-bromanyl-4,6-bis chloranyl aniline | |
C1=C(C=C(C(=C1Cl)N)Br)Cl | |
240.909 | |
240.92 | |
2-Bromo-4,6-dichloroaniline |
Safety and Handling
GHS H Statement
H301-H311-H332-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only