missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4,6-dinitroaniline, 97%, Thermo Scientific™

Product Code. 11419957
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1817-73-8
Molecular Formula C6H4BrN3O4
Molecular Weight (g/mol) 262.019
MDL Number MFCD00007146
InChI Key KWMDHCLJYMVBNS-UHFFFAOYSA-N
Synonym 6-bromo-2,4-dinitroaniline, bromo dna, 2,4-dinitro-6-bromoaniline, benzenamine, 2-bromo-4,6-dinitro, 2-bromo-4,6-dinitroaminobenzene, 2,4-dinitro-6-bromanilin, aniline, 2,4-dinitro-6-bromo, aniline, 2-bromo-4,6-dinitro, ccris 820, 2,4-dinitro-6-bromanilin czech
PubChem CID 15752
IUPAC Name 2-bromo-4,6-dinitroaniline
SMILES C1=C(C=C(C(=C1[N+](=O)[O-])N)Br)[N+](=O)[O-]

Specifications

Melting Point 151°C to 153°C
Quantity 25 g
Formula Weight 262.02
Percent Purity 97%
Chemical Name or Material 2-Bromo-4,6-dinitroaniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'