missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-4'-chloroacetophenone, 98%, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
100 g
500 g
Description
2-Bromo-4'-chloroacetophenone is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Bromo-4′-chloroacetophenone is used as pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
2-Bromo-4′-chloroacetophenone is used as pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
Specifications
Specifications
Melting Point | 94°C to 98°C |
Quantity | 100 g |
UN Number | UN3261 |
Beilstein | 607603 |
Merck Index | 14,2153 |
Solubility Information | Insoluble in water. |
Formula Weight | 233.5 |
Percent Purity | 98% |
Chemical Name or Material | 2-Bromo-4'-chloroacetophenone |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'