missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-fluorobenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Product Code. 11325555
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2-Bromo-4-fluorobenzenesulfonyl chloride is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-4-fluorobenzenesulfonyl chloride is used as a pharmaceutical intermediate.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive. Store away from oxidizing agents, water/ moisture, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 351003-45-7
Molecular Formula C6H3BrClFO2S
Molecular Weight (g/mol) 273.5
MDL Number MFCD03094149
InChI Key GJPWPFVLPNTOOL-UHFFFAOYSA-N
Synonym 2-bromo-4-fluorobenzene-1-sulfonyl chloride, 2-bromo-4-fluorobenzenesulphonyl chloride, 2-bromo-4-fluorobenzenesulfonylchloride, 4-fluoro-2-bromobenzenesulfonyl chloride, 2-bromo-4-fluorophenyl chlorosulfone, benzenesulfonyl chloride, 2-bromo-4-fluoro, pubchem8367, acmc-209vnu, 2-bromo-4-fluoro-benzenesulfonyl chloride, ksc221s9l
PubChem CID 2778216
IUPAC Name 2-bromo-4-fluorobenzenesulfonyl chloride
SMILES C1=CC(=C(C=C1F)Br)S(=O)(=O)Cl

Specifications

Density 1.87
Boiling Point 264°C to 265°C
Refractive Index 1.578
Quantity 1 g
UN Number UN2920
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes in water.
Formula Weight 273.51
Percent Purity 97%
Chemical Name or Material 2-Bromo-4-fluorobenzenesulfonyl chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.