missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-4-fluorobenzoic acid, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
2-Bromo-4-fluorobenzoic acid is used in the synthesis of 3-fluoro-8-(methylthio)dibenzo[b,f]thiepin-10(11H)-one, 2-fluoro-8-(methylthio)dibenzo[b,f]thiepin-10(11H)-one, 2-((2-carboxy-5-fluorophenyl)amino)-3-methoxybenzoic acid and 2-bromo-4-fluorobenzamide. It is also used in the preparation of boron-containing anti-fungal agents, in the treatment of onychomycosis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Bromo-4-fluorobenzoic acid is used in the synthesis of 3-fluoro-8-(methylthio)dibenzo[b,f]thiepin-10(11H)-one, 2-fluoro-8-(methylthio)dibenzo[b,f]thiepin-10(11H)-one, 2-((2-carboxy-5-fluorophenyl)amino)-3-methoxybenzoic acid and 2-bromo-4-fluorobenzamide. It is also used in the preparation of boron-containing anti-fungal agents, in the treatment of onychomycosis.
Solubility
Soluble in water and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
2-Bromo-4-fluorobenzoic acid is used in the synthesis of 3-fluoro-8-(methylthio)dibenzo[b,f]thiepin-10(11H)-one, 2-fluoro-8-(methylthio)dibenzo[b,f]thiepin-10(11H)-one, 2-((2-carboxy-5-fluorophenyl)amino)-3-methoxybenzoic acid and 2-bromo-4-fluorobenzamide. It is also used in the preparation of boron-containing anti-fungal agents, in the treatment of onychomycosis.
Solubility
Soluble in water and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 167°C to 169°C |
Quantity | 25 g |
Beilstein | 2614292 |
Solubility Information | Soluble in water and methanol. |
Formula Weight | 219.01 |
Percent Purity | 98% |
Chemical Name or Material | 2-Bromo-4-fluorobenzoic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'