Learn More
2-Bromo-4-fluorobenzoic acid, 98%, Thermo Scientific Chemicals
£20.70 - £151.00
Chemical Identifiers
CAS | 1006-41-3 |
---|---|
Molecular Formula | C7H4BrFO2 |
Molecular Weight (g/mol) | 219.009 |
MDL Number | MFCD00055370 |
InChI Key | RRKPMLZRLKTDQV-UHFFFAOYSA-N |
Synonym | 2-bromo-4-fluorobenzoicacid, 4-fluoro-2-bromobenzoic acid, 2-bromo-4-fluoro-benzoic acid, benzoic acid, 2-bromo-4-fluoro, rarechem al bo 1996, buttpark 18\02-85, 2-bromo-4-fluorbenzoic acid, pubchem1314, 2-bromo-4-fluorobenzoic, 2-bromofluorobenzoic acid |
PubChem CID | 1268169 |
IUPAC Name | 2-bromo-4-fluorobenzoic acid |
SMILES | C1=CC(=C(C=C1F)Br)C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11364236
|
Thermo Scientific Alfa Aesar
L19321.03 |
1 g |
£20.70
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11374236
|
Thermo Scientific Alfa Aesar
L19321.06 |
5 g |
£48.30
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11384236
|
Thermo Scientific Alfa Aesar
L19321.14 |
25 g |
£151.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2-Bromo-4-fluorobenzoic acid is used in the synthesis of 3-fluoro-8-(methylthio)dibenzo[b,f]thiepin-10(11H)-one, 2-fluoro-8-(methylthio)dibenzo[b,f]thiepin-10(11H)-one, 2-((2-carboxy-5-fluorophenyl)amino)-3-methoxybenzoic acid and 2-bromo-4-fluorobenzamide. It is also used in the preparation of boron-containing anti-fungal agents, in the treatment of onychomycosis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Bromo-4-fluorobenzoic acid is used in the synthesis of 3-fluoro-8-(methylthio)dibenzo[b,f]thiepin-10(11H)-one, 2-fluoro-8-(methylthio)dibenzo[b,f]thiepin-10(11H)-one, 2-((2-carboxy-5-fluorophenyl)amino)-3-methoxybenzoic acid and 2-bromo-4-fluorobenzamide. It is also used in the preparation of boron-containing anti-fungal agents, in the treatment of onychomycosis.
Solubility
Soluble in water and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Chemical Identifiers
1006-41-3 | |
219.009 | |
RRKPMLZRLKTDQV-UHFFFAOYSA-N | |
1268169 | |
C1=CC(=C(C=C1F)Br)C(=O)O |
C7H4BrFO2 | |
MFCD00055370 | |
2-bromo-4-fluorobenzoicacid, 4-fluoro-2-bromobenzoic acid, 2-bromo-4-fluoro-benzoic acid, benzoic acid, 2-bromo-4-fluoro, rarechem al bo 1996, buttpark 18\02-85, 2-bromo-4-fluorbenzoic acid, pubchem1314, 2-bromo-4-fluorobenzoic, 2-bromofluorobenzoic acid | |
2-bromo-4-fluorobenzoic acid |
Specifications
1006-41-3 | |
C7H4BrFO2 | |
2614292 | |
Soluble in water and methanol. | |
C1=CC(=C(C=C1F)Br)C(=O)O | |
219.009 | |
219.01 | |
2-Bromo-4-fluorobenzoic acid |
167°C to 169°C | |
MFCD00055370 | |
2-bromo-4-fluorobenzoicacid, 4-fluoro-2-bromobenzoic acid, 2-bromo-4-fluoro-benzoic acid, benzoic acid, 2-bromo-4-fluoro, rarechem al bo 1996, buttpark 18\02-85, 2-bromo-4-fluorbenzoic acid, pubchem1314, 2-bromo-4-fluorobenzoic, 2-bromofluorobenzoic acid | |
RRKPMLZRLKTDQV-UHFFFAOYSA-N | |
2-bromo-4-fluorobenzoic acid | |
1268169 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only