missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-fluorobenzyl alcohol, 98%, Thermo Scientific™

Product Code. 15438438
Click to view available options
Quantity:
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Bromo-4-fluorobenzyl alcohol, is used as an alcohol, medicine intermediate.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 229027-89-8
Molecular Formula C7H6BrFO
Molecular Weight (g/mol) 205.03
MDL Number MFCD00672925
InChI Key XGAMLBPEVCLQEJ-UHFFFAOYSA-N
PubChem CID 2773347
IUPAC Name (2-bromo-4-fluorophenyl)methanol
SMILES OCC1=C(Br)C=C(F)C=C1

Specifications

Melting Point 68°C to 72°C
Color White
Quantity 100 g
Synonym 2-bromo-4-fluorophenyl methanol, 2-bromo-4-fluorobenzyl alcohol, benzenemethanol, 2-bromo-4-fluoro, 2-bromo-4-fluorophenyl methan-1-ol, 2-bromo-4-fluorobenzenemethanol, benzenemethanol,2-bromo-4-fluoro, pubchem3222, acmc-1cqar, ksc550i1h, rarechem al bd 1088
Solubility Information Soluble in water.
Formula Weight 205.02
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Bromo-4-fluorobenzyl alcohol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'