missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-4-fluorobenzyl alcohol, 98%, Thermo Scientific™
£98.45
Chemical Identifiers
CAS | 229027-89-8 |
---|---|
Molecular Formula | C7H6BrFO |
Molecular Weight (g/mol) | 205.03 |
MDL Number | MFCD00672925 |
InChI Key | XGAMLBPEVCLQEJ-UHFFFAOYSA-N |
PubChem CID | 2773347 |
IUPAC Name | (2-bromo-4-fluorophenyl)methanol |
SMILES | OCC1=C(Br)C=C(F)C=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15438438
|
Thermo Scientific Alfa Aesar
H61850.22 |
100 g | |||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications2-Bromo-4-fluorobenzyl alcohol, is used as an alcohol, medicine intermediate.
Solubility
Soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
229027-89-8 | |
205.03 | |
XGAMLBPEVCLQEJ-UHFFFAOYSA-N | |
(2-bromo-4-fluorophenyl)methanol |
C7H6BrFO | |
MFCD00672925 | |
2773347 | |
OCC1=C(Br)C=C(F)C=C1 |
Specifications
229027-89-8 | |
White | |
MFCD00672925 | |
Soluble in water. | |
OCC1=C(Br)C=C(F)C=C1 | |
205.03 | |
205.02 | |
Solid |
68°C to 72°C | |
C7H6BrFO | |
2-bromo-4-fluorophenyl methanol, 2-bromo-4-fluorobenzyl alcohol, benzenemethanol, 2-bromo-4-fluoro, 2-bromo-4-fluorophenyl methan-1-ol, 2-bromo-4-fluorobenzenemethanol, benzenemethanol,2-bromo-4-fluoro, pubchem3222, acmc-1cqar, ksc550i1h, rarechem al bd 1088 | |
XGAMLBPEVCLQEJ-UHFFFAOYSA-N | |
(2-bromo-4-fluorophenyl)methanol | |
2773347 | |
98% | |
2-Bromo-4-fluorobenzyl alcohol |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures