missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-methoxyphenylacetic acid, 97%, Thermo Scientific Chemicals

Product Code. 11317469
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66916-99-2
Molecular Formula C9H9BrO3
Molecular Weight (g/mol) 245.07
MDL Number MFCD02664700
InChI Key XQELSBAAFMYSMG-UHFFFAOYSA-N
Synonym 2-bromo-4-methoxyphenylacetic acid, 2-2-bromo-4-methoxyphenyl acetic acid, 2-bromo-4-methoxyphenyl acetic acid, benzeneacetic acid, 2-bromo-4-methoxy, 2-bromo-4-methoxyphenylacetci acid, pubchem3983, 2-2-bromo-4-methoxy-phenyl acetic acid, benzeneacetic acid,2-bromo-4-methoxy, 2-bromo-4-methoxy-phenyl-acetic acid
PubChem CID 3963668
IUPAC Name 2-(2-bromo-4-methoxyphenyl)acetic acid
SMILES COC1=CC(Br)=C(CC(O)=O)C=C1

Specifications

Melting Point 127°C to 131°C
Quantity 25 g
Formula Weight 245.07
Percent Purity 97%
Chemical Name or Material 2-Bromo-4-methoxyphenylacetic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'