missing translation for 'onlineSavingsMsg'
Learn More
2-Bromo-4'-methylpropiophenone, 98%, Thermo Scientific Chemicals Product Code.: 15442279

2-Bromo-4'-methylpropiophenone, 98%, Thermo Scientific Chemicals

Product Code. 15442279
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 15442279

Brand: Thermo Scientific Alfa Aesar H64881.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field. Also used as intermediate for 4-methylmethcathinone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field. Also used as intermediate for 4-methylmethcathinone.

Solubility
Soluble in chloroform and methanol.

Notes
Store at 4°C. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1451-82-7
Molecular Formula C10H11BrO
Molecular Weight (g/mol) 227.10
MDL Number MFCD11131402
InChI Key OZLUPIIIHOOPNQ-UHFFFAOYNA-N
Synonym 2-bromo-1-p-tolyl propan-1-one, 2-bromo-4'-methylpropiophenone, 2-bromo-1-4-methylphenyl propan-1-one, 2-bromo-4-methylpropiophenone, 2-bromo-1-4-methylphenyl-1-propanone, 1-propanone, 2-bromo-1-4-methylphenyl, 2-bromo-1-p-tolylpropan-1-one, ksc917e0p, 2-bromo-1-p-tolyl-propan-1-one, 4'-methyl-alpha-bromopropiophenone
PubChem CID 2734063
IUPAC Name 2-bromo-1-(4-methylphenyl)propan-1-one
SMILES CC(Br)C(=O)C1=CC=C(C)C=C1

Specifications

Quantity 1 g
Beilstein 2207628
Solubility Information Soluble in chloroform and methanol.
Formula Weight 227.1
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Bromo-4'-methylpropiophenone

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM24E039 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberH64881.06,H64881.03,H64881.14
Lot Number81303035 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH64881.06,H64881.03,H64881.14
Lot Number81401386 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH64881.06,H64881.03,H64881.14
Lot NumberQ20E012 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberH64881.06,H64881.03,H64881.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Bromo-4'-methylpropiophenone, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.