missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4'-methylpropiophenone, 98%, Thermo Scientific Chemicals

Product Code. 15452279
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field. Also used as intermediate for 4-methylmethcathinone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field. Also used as intermediate for 4-methylmethcathinone.

Solubility
Soluble in chloroform and methanol.

Notes
Store at 4°C. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1451-82-7
Molecular Formula C10H11BrO
Molecular Weight (g/mol) 227.10
MDL Number MFCD11131402
InChI Key OZLUPIIIHOOPNQ-UHFFFAOYNA-N
Synonym 2-bromo-1-p-tolyl propan-1-one, 2-bromo-4'-methylpropiophenone, 2-bromo-1-4-methylphenyl propan-1-one, 2-bromo-4-methylpropiophenone, 2-bromo-1-4-methylphenyl-1-propanone, 1-propanone, 2-bromo-1-4-methylphenyl, 2-bromo-1-p-tolylpropan-1-one, ksc917e0p, 2-bromo-1-p-tolyl-propan-1-one, 4'-methyl-alpha-bromopropiophenone
PubChem CID 2734063
IUPAC Name 2-bromo-1-(4-methylphenyl)propan-1-one
SMILES CC(Br)C(=O)C1=CC=C(C)C=C1

Specifications

Quantity 5 g
Beilstein 2207628
Solubility Information Soluble in chloroform and methanol.
Formula Weight 227.1
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Bromo-4'-methylpropiophenone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'