missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-methylpyrimidine, 97%, Thermo Scientific Chemicals

£57.50 - £888.00

Chemical Identifiers

CAS 130645-48-6
Molecular Formula C5H5BrN2
Molecular Weight (g/mol) 173.013
MDL Number MFCD04037883
InChI Key UZIJEBOLOXOVFY-UHFFFAOYSA-N
PubChem CID 10942928
IUPAC Name 2-bromo-4-methylpyrimidine
SMILES CC1=NC(=NC=C1)Br
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15430788
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H54503.03
1 g
£57.50
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15440788
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H54503.06
5 g
£206.00
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15450788
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H54503.14
25 g
£888.00
25g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

2-Bromo-4-methylpyrimidine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-4-methylpyrimidine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

130645-48-6
173.013
UZIJEBOLOXOVFY-UHFFFAOYSA-N
2-bromo-4-methylpyrimidine
C5H5BrN2
MFCD04037883
10942928
CC1=NC(=NC=C1)Br

Specifications

130645-48-6
C5H5BrN2
2-bromo-4-methyl-pyrimidine, pyrimidine, 2-bromo-4-methyl, pyrimidine,2-bromo-4-methyl, pubchem7007, acmc-209z1w, 2-bromo-4-methlypyrimidine, 2-bromo-4-methyl pyrimidine, 2-bromo-4-methylpyrimidine
UZIJEBOLOXOVFY-UHFFFAOYSA-N
2-bromo-4-methylpyrimidine
10942928
97%
45°C to 50°C
MFCD04037883
Slightly soluble in water.
CC1=NC(=NC=C1)Br
173.013
173.01
2-Bromo-4-methylpyrimidine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only