missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4'-nitroacetophenone, 95%, Thermo Scientific Chemicals

Product Code. 10677441
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-81-0
Molecular Formula C8H6BrNO3
Molecular Weight (g/mol) 244.04
MDL Number MFCD00007356
InChI Key MBUPVGIGAMCMBT-UHFFFAOYSA-N
Synonym 2-bromo-4'-nitroacetophenone, 2-bromo-1-4-nitrophenyl ethanone, 4-nitrophenacyl bromide, p-nitrophenacyl bromide, 2-bromo-1-4-nitrophenyl ethan-1-one, alpha-bromo-p-nitroacetophenone, ethanone, 2-bromo-1-4-nitrophenyl, 4'-nitrophenacyl bromide, alpha-bromo-4-nitroacetophenone, 2-bromo-4-nitroacetophenone
PubChem CID 66840
IUPAC Name 2-bromo-1-(4-nitrophenyl)ethanone
SMILES C1=CC(=CC=C1C(=O)CBr)[N+](=O)[O-]

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 94°C to 100°C
Color Yellow to Orange
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula O2NC6H4COCH2Br
Quantity 10 g
Formula Weight 244.04
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-4′-nitroacetophenone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'