Learn More
2-Bromo-5-fluoro-3-nitropyridine, 95%, Thermo Scientific™
£63.65 - £190.00
Chemical Identifiers
CAS | 652160-72-0 |
---|---|
Molecular Formula | C5H2BrFN2O2 |
Molecular Weight (g/mol) | 220.99 |
MDL Number | MFCD05662387 |
InChI Key | XJFDIIHIXNIWQH-UHFFFAOYSA-N |
Synonym | 2-bromo-3-nitro-5-fluoropyridine, 2-bromo-5-fluoro-2-nitropyrdine, 2-bromo-5-fluoro-3-nitro-pyridine, pyridine,2-bromo-5-fluoro-3-nitro, pyridine, 2-bromo-5-fluoro-3-nitro, pubchem14284, 2-bromo-3-nitro-5-fluoro pyridine, 2-bromanyl-5-fluoranyl-3-nitro-pyridine |
PubChem CID | 20791243 |
IUPAC Name | 2-bromo-5-fluoro-3-nitropyridine |
SMILES | [O-][N+](=O)C1=CC(F)=CN=C1Br |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsIt is used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
652160-72-0 | |
220.99 | |
XJFDIIHIXNIWQH-UHFFFAOYSA-N | |
20791243 | |
[O-][N+](=O)C1=CC(F)=CN=C1Br |
C5H2BrFN2O2 | |
MFCD05662387 | |
2-bromo-3-nitro-5-fluoropyridine, 2-bromo-5-fluoro-2-nitropyrdine, 2-bromo-5-fluoro-3-nitro-pyridine, pyridine,2-bromo-5-fluoro-3-nitro, pyridine, 2-bromo-5-fluoro-3-nitro, pubchem14284, 2-bromo-3-nitro-5-fluoro pyridine, 2-bromanyl-5-fluoranyl-3-nitro-pyridine | |
2-bromo-5-fluoro-3-nitropyridine |
Specifications
652160-72-0 | |
C5H2BrFN2O2 | |
14253146 | |
Slightly soluble in water. | |
[O-][N+](=O)C1=CC(F)=CN=C1Br | |
220.99 | |
220.99 | |
Solid |
Orange | |
MFCD05662387 | |
2-bromo-3-nitro-5-fluoropyridine, 2-bromo-5-fluoro-2-nitropyrdine, 2-bromo-5-fluoro-3-nitro-pyridine, pyridine,2-bromo-5-fluoro-3-nitro, pyridine, 2-bromo-5-fluoro-3-nitro, pubchem14284, 2-bromo-3-nitro-5-fluoro pyridine, 2-bromanyl-5-fluoranyl-3-nitro-pyridine | |
XJFDIIHIXNIWQH-UHFFFAOYSA-N | |
2-bromo-5-fluoro-3-nitropyridine | |
20791243 | |
95% | |
2-Bromo-5-fluoro-3-nitropyridine |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures