missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-fluoro-3-nitropyridine, 95%, Thermo Scientific™

Product Code. 15461309
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 652160-72-0
Molecular Formula C5H2BrFN2O2
Molecular Weight (g/mol) 220.99
MDL Number MFCD05662387
InChI Key XJFDIIHIXNIWQH-UHFFFAOYSA-N
Synonym 2-bromo-3-nitro-5-fluoropyridine, 2-bromo-5-fluoro-2-nitropyrdine, 2-bromo-5-fluoro-3-nitro-pyridine, pyridine,2-bromo-5-fluoro-3-nitro, pyridine, 2-bromo-5-fluoro-3-nitro, pubchem14284, 2-bromo-3-nitro-5-fluoro pyridine, 2-bromanyl-5-fluoranyl-3-nitro-pyridine
PubChem CID 20791243
IUPAC Name 2-bromo-5-fluoro-3-nitropyridine
SMILES [O-][N+](=O)C1=CC(F)=CN=C1Br

Specifications

Color Orange
Quantity 250 mg
Beilstein 14253146
Solubility Information Slightly soluble in water.
Formula Weight 220.99
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Bromo-5-fluoro-3-nitropyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'