Learn More
2-Bromo-5-fluorotoluene, 98+%, Thermo Scientific Chemicals
£39.40 - £128.64
Chemical Identifiers
CAS | 452-63-1 |
---|---|
Molecular Formula | C7H6BrF |
Molecular Weight (g/mol) | 189.027 |
MDL Number | MFCD00017921 |
InChI Key | RJPNVPITBYXBNB-UHFFFAOYSA-N |
Synonym | 2-bromo-5-fluorotoluene, 3-fluoro-6-bromotoluene, 2-bromo-5-fluoro toluene, benzene, 1-bromo-4-fluoro-2-methyl, 5-fluoro-2-bromotoluene, 1-bromo-4-fluoro-2-methyl-benzene, pubchem1603, 2-bromo-5fluorotoluene, acmc-1acnn, 2-bromo-5-fluoro-toluene |
PubChem CID | 96743 |
IUPAC Name | 1-bromo-4-fluoro-2-methylbenzene |
SMILES | CC1=C(C=CC(=C1)F)Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11478993
|
Thermo Scientific Alfa Aesar
A11112.14 |
25 g |
£39.40
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11488993
|
Thermo Scientific Alfa Aesar
A11112.22 |
100 g |
£128.64
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2-Bromo-5-fluorotoluene is used to produce other chemicals. It can produce 2-Bromo-5-fluoro-benzoic acid. The reaction occurs with reagent KMnO4, solvent H2O and other condition of heating.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Bromo-5-fluorotoluene is used to produce other chemicals. It can produce 2-Bromo-5-fluoro-benzoic acid. The reaction occurs with reagent KMnO4, solvent H2O and other condition of heating.
Solubility
Difficult to mix.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
452-63-1 | |
189.027 | |
RJPNVPITBYXBNB-UHFFFAOYSA-N | |
96743 | |
CC1=C(C=CC(=C1)F)Br |
C7H6BrF | |
MFCD00017921 | |
2-bromo-5-fluorotoluene, 3-fluoro-6-bromotoluene, 2-bromo-5-fluoro toluene, benzene, 1-bromo-4-fluoro-2-methyl, 5-fluoro-2-bromotoluene, 1-bromo-4-fluoro-2-methyl-benzene, pubchem1603, 2-bromo-5fluorotoluene, acmc-1acnn, 2-bromo-5-fluoro-toluene | |
1-bromo-4-fluoro-2-methylbenzene |
Specifications
452-63-1 | |
169°C to 170°C | |
C7H6BrF | |
MFCD00017921 | |
2-bromo-5-fluorotoluene, 3-fluoro-6-bromotoluene, 2-bromo-5-fluoro toluene, benzene, 1-bromo-4-fluoro-2-methyl, 5-fluoro-2-bromotoluene, 1-bromo-4-fluoro-2-methyl-benzene, pubchem1603, 2-bromo-5fluorotoluene, acmc-1acnn, 2-bromo-5-fluoro-toluene | |
RJPNVPITBYXBNB-UHFFFAOYSA-N | |
1-bromo-4-fluoro-2-methylbenzene | |
96743 | |
≥98% |
1.495 | |
70°C (158°F) | |
1.526 | |
1859121 | |
Difficult to mix. | |
CC1=C(C=CC(=C1)F)Br | |
189.027 | |
189.03 | |
2-Bromo-5-fluorotoluene |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
missing translation for 'dotInformation' : Hazard Class: 3; Packaging Group: III
missing translation for 'einecsNumber' : 207-203-5
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.