missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-hydroxypyrazine, 97%, Thermo Scientific Chemicals

£221.00 - £424.00

Chemical Identifiers

CAS 374063-92-0
Molecular Formula C4H3BrN2O
Molecular Weight (g/mol) 174.99
MDL Number MFCD06245330
InChI Key ITTXBHQAWOFJAI-UHFFFAOYSA-N
Synonym 5-bromopyrazin-2-ol, 2-bromo-5-hydroxypyrazine, 5-bromo-2-pyrazinol, 5-bromo-pyrazin-2-ol, 2 1h-pyrazinone, 5-bromo, 5-bromo-2-hydroxypyrazine, 2-bromo-5-hydroxy-pyrazine, pubchem7870, acmc-20a0of, 2-hydroxy-5-bromopyrazine
PubChem CID 2771618
SMILES BrC1=CNC(=O)C=N1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10586459
View Documents
Thermo Scientific Acros
435890010
1 g Glass bottle
£221.00
1g
Please to purchase this item. Need a web account? Register with us today!
10194069
View Documents
Thermo Scientific Acros
435890050
5 g Glass bottle
£424.00
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

374063-92-0
174.99
ITTXBHQAWOFJAI-UHFFFAOYSA-N
2771618
C4H3BrN2O
MFCD06245330
5-bromopyrazin-2-ol, 2-bromo-5-hydroxypyrazine, 5-bromo-2-pyrazinol, 5-bromo-pyrazin-2-ol, 2 1h-pyrazinone, 5-bromo, 5-bromo-2-hydroxypyrazine, 2-bromo-5-hydroxy-pyrazine, pubchem7870, acmc-20a0of, 2-hydroxy-5-bromopyrazine
BrC1=CNC(=O)C=N1

Specifications

374063-92-0
100.0
White to Brown
96% min. (HPLC)
MFCD06245330
ITTXBHQAWOFJAI-UHFFFAOYSA-N
5-bromo-1,2-dihydropyrazin-2-one
2771618
97%
2-Bromo-5-hydroxypyrazine
96.0
115°C to 119°C
Authentic
C4H3BrN2O
5-bromopyrazin-2-ol, 2-bromo-5-hydroxypyrazine, 5-bromo-2-pyrazinol, 5-bromo-pyrazin-2-ol, 2 1h-pyrazinone, 5-bromo, 5-bromo-2-hydroxypyrazine, 2-bromo-5-hydroxy-pyrazine, pubchem7870, acmc-20a0of, 2-hydroxy-5-bromopyrazine
BrC1=CNC(=O)C=N1
174.99
174.98
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap

GHS Signal Word: Warning

Product Identifier
  • 2-Bromo-5-hydroxypyrazine

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only