missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-methoxybenzeneboronic acid, 97%, Thermo Scientific™

Product Code. 15469737
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89694-44-0
Molecular Formula C7H8BBrO3
Molecular Weight (g/mol) 230.852
MDL Number MFCD06659859
InChI Key BJQKEDXKQVNQPR-UHFFFAOYSA-N
Synonym 2-bromo-5-methoxybenzene boronic acid, 2-bromo-5-methoxyphenyl boronic acid, 2-bromo-5-methoxy benzeneboronic acid, acmc-209r2f, amtb841, 2-bromo-5-methoxyphenylboranic acid, 2-bromo-5-methoxyphenylboronicacid, boronic acid,b-2-bromo-5-methoxyphenyl
PubChem CID 25266062
IUPAC Name (2-bromo-5-methoxyphenyl)boronic acid
SMILES B(C1=C(C=CC(=C1)OC)Br)(O)O

Specifications

Melting Point 214°C to 216°C
Quantity 1 g
Formula Weight 230.85
Percent Purity 97%
Chemical Name or Material 2-Bromo-5-methoxybenzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'