missing translation for 'onlineSavingsMsg'
Learn More
2-Bromo-5-methoxybenzeneboronic acid, 97%, Thermo Scientific™ Product Code.: 15479737

2-Bromo-5-methoxybenzeneboronic acid, 97%, Thermo Scientific™

Product Code. 15479737
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15479737

Brand: Thermo Scientific Alfa Aesar H52972.MD

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89694-44-0
Molecular Formula C7H8BBrO3
Molecular Weight (g/mol) 230.852
MDL Number MFCD06659859
InChI Key BJQKEDXKQVNQPR-UHFFFAOYSA-N
Synonym 2-bromo-5-methoxybenzene boronic acid, 2-bromo-5-methoxyphenyl boronic acid, 2-bromo-5-methoxy benzeneboronic acid, acmc-209r2f, amtb841, 2-bromo-5-methoxyphenylboranic acid, 2-bromo-5-methoxyphenylboronicacid, boronic acid,b-2-bromo-5-methoxyphenylShow MoreShow Less
PubChem CID 25266062
IUPAC Name (2-bromo-5-methoxyphenyl)boronic acid
SMILES B(C1=C(C=CC(=C1)OC)Br)(O)O

Specifications

Melting Point 214°C to 216°C
Quantity 250 mg
Formula Weight 230.85
Percent Purity 97%
Chemical Name or Material 2-Bromo-5-methoxybenzeneboronic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberB-BH080502 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Bromo-5-methoxybenzeneboronic acid, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.