Learn More
2-Bromo-5-methoxybenzyl bromide, 97%, Thermo Scientific Chemicals
£35.10 - £143.00
Chemical Identifiers
CAS | 19614-12-1 |
---|---|
Molecular Formula | C8H8Br2O |
Molecular Weight (g/mol) | 279.959 |
MDL Number | MFCD00078664 |
InChI Key | MURVUTUZSUEIGI-UHFFFAOYSA-N |
Synonym | 2-bromo-5-methoxybenzyl bromide, 1-bromo-2-bromomethyl-4-methoxybenzene, 2-bromo-5-methoxybenzylbromide, 4-bromo-3-bromomethyl anisole, 2-bromo-5-methoxy-benzyl bromide, benzene, 1-bromo-2-bromomethyl-4-methoxy, 4-bromo-3-bromomethyl-1-methoxybenzene, pubchem18176, acmc-20a5x2, 4-bromo3-bromomethylanisole |
PubChem CID | 3525951 |
IUPAC Name | 1-bromo-2-(bromomethyl)-4-methoxybenzene |
SMILES | COC1=CC(=C(C=C1)Br)CBr |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15452399
|
Thermo Scientific Alfa Aesar
H64963.06 |
5 g |
£35.10
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15462399
|
Thermo Scientific Alfa Aesar
H64963.14 |
25 g |
£143.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEmployed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field
Solubility
Soluble in hot methanol. Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Chemical Identifiers
19614-12-1 | |
279.959 | |
MURVUTUZSUEIGI-UHFFFAOYSA-N | |
3525951 | |
COC1=CC(=C(C=C1)Br)CBr |
C8H8Br2O | |
MFCD00078664 | |
2-bromo-5-methoxybenzyl bromide, 1-bromo-2-bromomethyl-4-methoxybenzene, 2-bromo-5-methoxybenzylbromide, 4-bromo-3-bromomethyl anisole, 2-bromo-5-methoxy-benzyl bromide, benzene, 1-bromo-2-bromomethyl-4-methoxy, 4-bromo-3-bromomethyl-1-methoxybenzene, pubchem18176, acmc-20a5x2, 4-bromo3-bromomethylanisole | |
1-bromo-2-(bromomethyl)-4-methoxybenzene |
Specifications
19614-12-1 | |
White | |
MFCD00078664 | |
1945118 | |
Soluble in hot methanol. Slightly soluble in water. | |
COC1=CC(=C(C=C1)Br)CBr | |
279.959 | |
279.96 | |
Solid |
90°C to 94°C (literature) | |
C8H8Br2O | |
UN3261 | |
2-bromo-5-methoxybenzyl bromide, 1-bromo-2-bromomethyl-4-methoxybenzene, 2-bromo-5-methoxybenzylbromide, 4-bromo-3-bromomethyl anisole, 2-bromo-5-methoxy-benzyl bromide, benzene, 1-bromo-2-bromomethyl-4-methoxy, 4-bromo-3-bromomethyl-1-methoxybenzene, pubchem18176, acmc-20a5x2, 4-bromo3-bromomethylanisole | |
MURVUTUZSUEIGI-UHFFFAOYSA-N | |
1-bromo-2-(bromomethyl)-4-methoxybenzene | |
3525951 | |
97% | |
2-Bromo-5-methoxybenzyl bromide |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H290-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only