missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-methylbenzonitrile, 97%, Thermo Scientific™

Product Code. 15458178
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42872-83-3
Molecular Formula C8H6BrN
Molecular Weight (g/mol) 196.05
MDL Number MFCD11040281
InChI Key AKCXJAVATJLYQM-UHFFFAOYSA-N
Synonym 6-bromo-m-tolunitrile, 4-bromo-3-cyanotoluene, benzonitrile,2-bromo-5-methyl, benzonitrile, 2-bromo-5-methyl, pubchem14563, acmc-1aktv, ksc494a0n, 2-bromo-5-methyl-benzonitrile, 2-bromo-5-methyl benzonitrile, 2-bromo-5-methylbenzonitrile
PubChem CID 12557201
IUPAC Name 2-bromo-5-methylbenzonitrile
SMILES CC1=CC(C#N)=C(Br)C=C1

Specifications

Melting Point 61°C to 65°C
Color White to Yellow
Quantity 1 g
UN Number UN3439
Formula Weight 196.04
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-5-methylbenzonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'