missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-nitrophenetole, 98%, Thermo Scientific™

Product Code. 15426527
Click to view available options
Quantity:
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 423165-33-7
Molecular Formula C8H8BrNO3
Molecular Weight (g/mol) 246.06
MDL Number MFCD03790890
InChI Key ZALXYXWTUXLPBJ-UHFFFAOYSA-N
Synonym 2-bromo-5-nitrophenetole, 2-bromo-5-nitrophenyl ethyl ether, benzene,1-bromo-2-ethoxy-4-nitro, pubchem3240, ksc586a2t, 2-ethoxy-4-nitrophenyl bromide
PubChem CID 22832075
IUPAC Name 1-bromo-2-ethoxy-4-nitrobenzene
SMILES CCOC1=C(C=CC(=C1)[N+](=O)[O-])Br

Specifications

Melting Point 52°C to 56°C
Quantity 25 g
Formula Weight 246.06
Percent Purity 98%
Chemical Name or Material 2-Bromo-5-nitrophenetole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'