missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-ethoxynaphthalene, 98%, Thermo Scientific™

Product Code. 11420040
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66217-19-4
Molecular Formula C12H11BrO
Molecular Weight (g/mol) 251.12
MDL Number MFCD01568865
InChI Key FXVYIQCXQULCKL-UHFFFAOYSA-N
PubChem CID 236858
IUPAC Name 2-bromo-6-ethoxynaphthalene
SMILES CCOC1=CC=C2C=C(Br)C=CC2=C1

Specifications

Quantity 1 g
Synonym 6-ethoxy-2-bromonaphthalene, naphthalene,2-bromo-6-ethoxy, 6-bromo-2-ethoxynaphthalene, acmc-209nu2, 2-bromo-6-ethoxy-naphthalene, 6-bromo-2-naphthyl ethyl ether, naphthalene, 2-bromo-6-ethoxy
Formula Weight 251.12
Percent Purity 98%
Chemical Name or Material 2-Bromo-6-ethoxynaphthalene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'