missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-nitrotoluene, 98%, Thermo Scientific Chemicals

Product Code. 11440227
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55289-35-5
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.034
MDL Number MFCD00009792
InChI Key LYTNSGFSAXWBCA-UHFFFAOYSA-N
Synonym 2-bromo-6-nitrotoluene, 6-bromo-2-nitrotoluene, 3-bromo-2-methylnitrobenzene, 1-bromo-2-methyl-3-nitro-benzene, benzene, 1-bromo-2-methyl-3-nitro, bromo 2--6-nitrotoluene, 2-bromo-6-nitrotoluene,2-nitro-6-bromotoluene, 3-bromo-2-methyl-1-nitrobenzene, 1-brom-2-methyl-3-nitrobenzol, pubchem3823
PubChem CID 123537
IUPAC Name 1-bromo-2-methyl-3-nitrobenzene
SMILES CC1=C(C=CC=C1Br)[N+](=O)[O-]

Specifications

Melting Point 37°C to 42°C
Boiling Point 108°C to 110°C (3.3 mmHg)
Flash Point >110°C (230°F)
Quantity 1 g
Beilstein 2502581
Formula Weight 216.04
Percent Purity 98%
Chemical Name or Material 2-Bromo-6-nitrotoluene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'