missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzene-1-sulfonamide, 97%, Thermo Scientific™

Product Code. 10213482
Click to view available options
Quantity:
1 g
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 92748-09-9
Molecular Formula C6H6BrNO2S
Molecular Weight (g/mol) 236.08
MDL Number MFCD00173662
InChI Key YSFGGXNLZUSHHS-UHFFFAOYSA-N
Synonym 2-bromobenzene-1-sulfonamide, 2-bromobenzenesulphonamide, 2-bromo-benzenesulfonamide, 2-bromobenzene sulfonamide, 2-bromo benzene sulfon amide, buttpark 89\07-69, 2-bromobenzene-1-sulphonamide, bromobenzenesulfonamide, bromobenzene sulfonamide, pubchem13521
PubChem CID 2801263
IUPAC Name 2-bromobenzenesulfonamide
SMILES NS(=O)(=O)C1=CC=CC=C1Br

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula BrC6H4SO2NH2
Quantity 1 g
Formula Weight 236.09
Percent Purity 97%
Chemical Name or Material 2-Bromobenzene-1-sulfonamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'