missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzeneboronic acid, 98%, Thermo Scientific Chemicals

Product Code. 11319425
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2-Bromobenzeneboronic acid is used to promote greener amidations of carboxylic acids and amines in catalytic amounts. This technology avoids the requirement of preactivation of the carboxylic acid or use of coupling reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromobenzeneboronic acid is used to promote greener amidations of carboxylic acids and amines in catalytic amounts. This technology avoids the requirement of preactivation of the carboxylic acid or use of coupling reagents.

Solubility
Soluble in methanol. Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 244205-40-1
Molecular Formula C6H6BBrO2
Molecular Weight (g/mol) 200.83
MDL Number MFCD01114672
InChI Key PLVCYMZAEQRYHJ-UHFFFAOYSA-N
Synonym 2-bromophenyl boronic acid, o-bromophenylboronic acid, 2-bromobenzeneboronic acid, 2-bromophenylboronicacid, boronic acid, 2-bromophenyl, bromobenzeneboronic acid, pubchem13095, 2-bromo phenyl boronic acid, 2-bromo-phenyl boronic acid
PubChem CID 2773294
IUPAC Name (2-bromophenyl)boronic acid
SMILES OB(O)C1=CC=CC=C1Br

Specifications

Melting Point 108°C to 110°C
Quantity 1 g
Beilstein 8542615
Solubility Information Soluble in methanol. Slightly soluble in water.
Formula Weight 200.83
Percent Purity 98%
Chemical Name or Material 2-Bromobenzeneboronic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.