missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzimidazole, 99%, Thermo Scientific Chemicals

£54.40 - £188.00

Chemical Identifiers

CAS 54624-57-6
Molecular Formula C7H5BrN2
Molecular Weight (g/mol) 197.035
MDL Number MFCD00160009
InChI Key PHPYXVIHDRDPDI-UHFFFAOYSA-N
PubChem CID 2776281
IUPAC Name 2-bromo-1H-benzimidazole
SMILES C1=CC=C2C(=C1)NC(=N2)Br
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11346579
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H26183.03
1 g
£54.40
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11356579
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H26183.06
5 g
£188.00
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Reactant in the synthesis of substituted benzimidazoles. 2-Bromobenzimidazole is the starting compound in the synthesis of polyhalogenobenzimidazoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant in the synthesis of substituted benzimidazoles. 2-Bromobenzimidazole is the starting compound in the synthesis of polyhalogenobenzimidazoles.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Chemical Identifiers

54624-57-6
197.035
PHPYXVIHDRDPDI-UHFFFAOYSA-N
2-bromo-1H-benzimidazole
C7H5BrN2
MFCD00160009
2776281
C1=CC=C2C(=C1)NC(=N2)Br

Specifications

54624-57-6
C7H5BrN2
2-bromobenzimidazole, 2-bromo-1h-benzo d imidazole, 2-bromo-1h-1,3-benzodiazole, 2-bromo-1h-benzoimidazole, 1h-benzimidazole, 2-bromo, bromobenzimidazole, bromo-benzimidazole, pubchem16113, acmc-1an7o, 1h-benzimidazole,2-bromo
PHPYXVIHDRDPDI-UHFFFAOYSA-N
2-bromo-1H-benzimidazole
2776281
99%
195°C to 201°C
MFCD00160009
Insoluble in water.
C1=CC=C2C(=C1)NC(=N2)Br
197.035
197.03
2-Bromobenzimidazole
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only