missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobiphenyl, 98%, Thermo Scientific Chemicals

Product Code. 10588463
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2052-07-5
Molecular Formula C12H9Br
Molecular Weight (g/mol) 233.11
MDL Number MFCD00000065
InChI Key KTADSLDAUJLZGL-UHFFFAOYSA-N
Synonym 2-bromobiphenyl, o-bromobiphenyl, 2-bromo-biphenyl, 1,1'-biphenyl, 2-bromo, 2-bromo-1,1'-biphenyl, biphenyl, 2-bromo, 2-bromo-diphenyl, bromobiphenyl, 2-bromodiphenyl, 1,1'-biphenyl, bromo
PubChem CID 16329
IUPAC Name 1-bromo-2-phenylbenzene
SMILES C1=CC=C(C=C1)C2=CC=CC=C2Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 1.5°C to 2.0°C
Color Colorless
Density 1.3520g/mL
Boiling Point 297.0°C to 298.0°C
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5C6H4Br
Refractive Index 1.6280 to 1.6300
Quantity 1 g
Specific Gravity 1.352
Formula Weight 233.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Bromobiphenyl
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromobiphenyl
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'