missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobutane, 99+%, Thermo Scientific Chemicals

£19.60 - £225.12

Chemical Identifiers

CAS 78-76-2
Molecular Formula C4H9Br
Molecular Weight (g/mol) 137.02
MDL Number MFCD00000156
InChI Key UPSXAPQYNGXVBF-UHFFFAOYSA-N
Synonym sec-butyl bromide, butane, 2-bromo, 2-butyl bromide, methylethylbromomethane, 2-bromo-butane, secondary butyl bromide, sec-butylbromide, 1-bromo-1-methylpropane, ccris 106, bromobutane, 2
PubChem CID 6554
IUPAC Name 2-bromobutane
SMILES CCC(C)Br
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10539990
View Documents
Thermo Scientific Acros
106781000
100 mL Glass bottle
£19.60
100mL
Please to purchase this item. Need a web account? Register with us today!
10325930
View Documents
Thermo Scientific Acros
106785000
500 mL Glass bottle
£55.48
500mL
Please to purchase this item. Need a web account? Register with us today!
10175593
View Documents Promotion Details
Thermo Scientific Acros
106780025
2.5 L Glass bottle
£225.12
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

78-76-2
137.02
UPSXAPQYNGXVBF-UHFFFAOYSA-N
6554
CCC(C)Br
C4H9Br
MFCD00000156
sec-butyl bromide, butane, 2-bromo, 2-butyl bromide, methylethylbromomethane, 2-bromo-butane, secondary butyl bromide, sec-butylbromide, 1-bromo-1-methylpropane, ccris 106, bromobutane, 2
2-bromobutane

Specifications

78-76-2
100.0
1.2500g/mL
21°C
99% min. (GC)
C2H5CH(Br)CH3
MFCD00000156
15,1556
sec-butyl bromide, butane, 2-bromo, 2-butyl bromide, methylethylbromomethane, 2-bromo-butane, secondary butyl bromide, sec-butylbromide, 1-bromo-1-methylpropane, ccris 106, bromobutane, 2
UPSXAPQYNGXVBF-UHFFFAOYSA-N
2-bromobutane
6554
99+%
2-Bromobutane, 99+%
99.0
-112°C
91°C
Authentic
C4H9Br
1.4359 to 1.4379
01,119
1.25
Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
CCC(C)Br
137.02
137.02
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful to aquatic life with long lasting effects.
Highly flammable liquid and vapor.

GHS P Statement
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 201-140-7

missing translation for 'rtecsNumber' : EJ6228000

missing translation for 'tsca' : TSCA

Product Identifier
  • 2-Bromobutane

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only