missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromodibenzofuran, 98%, Thermo Scientific Chemicals
£34.60 - £438.00
Chemical Identifiers
CAS | 86-76-0 |
---|---|
Molecular Formula | C12H7BrO |
Molecular Weight (g/mol) | 247.091 |
MDL Number | MFCD00092338 |
InChI Key | CRJISNQTZDMKQD-UHFFFAOYSA-N |
Synonym | 2-bromodibenzo b,d furan, 2-bromo-dibenzofuran, dibenzofuran, 2-bromo, dibenzofuran, 2-bromo-8ci 9ci, 2-bromodibenzo b furan, 4-bromo-8-oxatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2,4,6,10,12-hexaene, 4-bromo-8-oxatricyclo 7.4.0.0 2 , trideca-1 9 ,2 7 ,3,5,10,12-hexaene, 4-bromo-8-oxatricyclo 7.4.0.0?,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene, 4-bromo-8-oxatricyclo 7.4.0.0^ 2,7 trideca-1 13 ,2,4,6,9,11-hexaene |
PubChem CID | 6856 |
IUPAC Name | 2-bromodibenzofuran |
SMILES | C1=CC=C2C(=C1)C3=C(O2)C=CC(=C3)Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15401879
|
Thermo Scientific Alfa Aesar
H64596.MD |
250 mg | N/A | ||||||
15481869
|
Thermo Scientific Alfa Aesar
H64596.03 |
1 g |
£108.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15491869
|
Thermo Scientific Alfa Aesar
H64596.06 |
5 g | N/A | ||||||
Description
2-Bromodibenzofuran is used as a reagent in Friedel-Crafts acylation reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Bromodibenzofuran is used as a reagent in Friedel-Crafts acylation reactions.
Solubility
Soluble in hot methanol.
Notes
Incompatible with oxidizing agents.
Chemical Identifiers
86-76-0 | |
247.091 | |
CRJISNQTZDMKQD-UHFFFAOYSA-N | |
6856 | |
C1=CC=C2C(=C1)C3=C(O2)C=CC(=C3)Br |
C12H7BrO | |
MFCD00092338 | |
2-bromodibenzo b,d furan, 2-bromo-dibenzofuran, dibenzofuran, 2-bromo, dibenzofuran, 2-bromo-8ci 9ci, 2-bromodibenzo b furan, 4-bromo-8-oxatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2,4,6,10,12-hexaene, 4-bromo-8-oxatricyclo 7.4.0.0 2 , trideca-1 9 ,2 7 ,3,5,10,12-hexaene, 4-bromo-8-oxatricyclo 7.4.0.0?,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene, 4-bromo-8-oxatricyclo 7.4.0.0^ 2,7 trideca-1 13 ,2,4,6,9,11-hexaene | |
2-bromodibenzofuran |
Specifications
86-76-0 | |
C12H7BrO | |
142199 | |
Soluble in hot methanol. | |
C1=CC=C2C(=C1)C3=C(O2)C=CC(=C3)Br | |
247.091 | |
247.09 | |
Solid |
108°C to 111°C | |
MFCD00092338 | |
2-bromodibenzo b,d furan, 2-bromo-dibenzofuran, dibenzofuran, 2-bromo, dibenzofuran, 2-bromo-8ci 9ci, 2-bromodibenzo b furan, 4-bromo-8-oxatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2,4,6,10,12-hexaene, 4-bromo-8-oxatricyclo 7.4.0.0 2 , trideca-1 9 ,2 7 ,3,5,10,12-hexaene, 4-bromo-8-oxatricyclo 7.4.0.0?,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene, 4-bromo-8-oxatricyclo 7.4.0.0^ 2,7 trideca-1 13 ,2,4,6,9,11-hexaene | |
CRJISNQTZDMKQD-UHFFFAOYSA-N | |
2-bromodibenzofuran | |
6856 | |
98% | |
2-Bromodibenzofuran |
Safety and Handling
GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.
P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
H302-H319
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only