Learn More
2-Bromoethyl acetate, 97%, Thermo Scientific Chemicals
£14.86 - £104.84
Chemical Identifiers
CAS | 927-68-4 |
---|---|
Molecular Formula | C4H7BrO2 |
Molecular Weight (g/mol) | 167.00 |
MDL Number | MFCD00000235 |
InChI Key | RGHQKFQZGLKBCF-UHFFFAOYSA-N |
Synonym | bromoethyl acetate, 1-acetoxy-2-bromoethane, 2-bromoethylacetate, ethanol, 2-bromo-, acetate, 1-bromo-2-acetoxyethane, acetic acid 2-bromoethyl ester, unii-nrz256o7qb, ethanol, 2-bromo-, 1-acetate, beta-bromoethyl acetate, nrz256o7qb |
PubChem CID | 13564 |
IUPAC Name | 2-bromoethyl acetate |
SMILES | CC(=O)OCCBr |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
10646270
|
Thermo Scientific Acros
160090050 |
5 g | Glass bottle | N/A | ||||||
10048139
|
Thermo Scientific Acros
160090250 |
25 g | Glass bottle | N/A | ||||||
10493340
|
Thermo Scientific Acros
160091000 |
100 g | Glass bottle | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
927-68-4 | |
167.00 | |
RGHQKFQZGLKBCF-UHFFFAOYSA-N | |
13564 | |
CC(=O)OCCBr |
C4H7BrO2 | |
MFCD00000235 | |
bromoethyl acetate, 1-acetoxy-2-bromoethane, 2-bromoethylacetate, ethanol, 2-bromo-, acetate, 1-bromo-2-acetoxyethane, acetic acid 2-bromoethyl ester, unii-nrz256o7qb, ethanol, 2-bromo-, 1-acetate, beta-bromoethyl acetate, nrz256o7qb | |
2-bromoethyl acetate |
Specifications
927-68-4 | |
100.0 | |
Colorless to Yellow | |
160.0°C to 162.0°C | |
Authentic | |
C4H7BrO2 | |
1.4547 | |
1.51 | |
RGHQKFQZGLKBCF-UHFFFAOYSA-N | |
2-bromoethyl acetate | |
13564 | |
97% | |
2-Bromoethyl acetate |
96.0 | |
-13.8°C | |
1.5100g/mL | |
71°C | |
96% min. (GC) | |
CH3CO2CH2CH2Br | |
MFCD00000235 | |
bromoethyl acetate, 1-acetoxy-2-bromoethane, 2-bromoethylacetate, ethanol, 2-bromo-, acetate, 1-bromo-2-acetoxyethane, acetic acid 2-bromoethyl ester, unii-nrz256o7qb, ethanol, 2-bromo-, 1-acetate, beta-bromoethyl acetate, nrz256o7qb | |
CC(=O)OCCBr | |
167.00 | |
167 | |
Liquid |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 213-159-8
- 2-Bromoethyl acetate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.