missing translation for 'onlineSavingsMsg'
Learn More

(2-Bromoethyl)benzene, 98%, Thermo Scientific Chemicals

Product Code. 11404194
100 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

(2-Bromoethyl)benzene is used in the preparation of phenelzine by reacting with hydrazine. It is also used as a starting material to prepare various beta-phenethyl derivatives, active pharmaceutical ingredients, and fragrances. It finds application as a flame retardant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(2-Bromoethyl)benzene is used in the preparation of phenelzine by reacting with hydrazine. It is also used as a starting material to prepare various beta-phenethyl derivatives, active pharmaceutical ingredients, and fragrances. It finds application as a flame retardant.

Solubility
Miscible with ether and benzene. Immiscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-63-9
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.064
MDL Number MFCD00000240
InChI Key WMPPDTMATNBGJN-UHFFFAOYSA-N
Synonym 2-bromoethyl benzene, phenethyl bromide, 2-phenylethyl bromide, 1-bromo-2-phenylethane, phenylethyl bromide, benzene, 2-bromoethyl, 2-phenethyl bromide, beta-bromoethylbenzene, 2-phenyl-1-bromoethane, phenethylbromide
PubChem CID 7666
IUPAC Name 2-bromoethylbenzene
SMILES C1=CC=C(C=C1)CCBr

Specifications

Melting Point -56°C
Density 1.355
Boiling Point 220°C to 221°C
Flash Point 89°C (192°F)
Linear Formula C6H5(CH2)2Br
Refractive Index 1.556
Quantity 100 g
Beilstein 507487
Solubility Information Miscible with ether and benzene. Immiscible with water.
Formula Weight 185.06
Percent Purity 98%
Chemical Name or Material (2-Bromoethyl)benzene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(2-Bromoethyl)benzene, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.