missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromomalonaldehyde, 97+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 132.0°C to 136.0°C |
Assay Percent Range | 98.5 % min. (Acid) |
Packaging | Glass bottle |
Linear Formula | BrCH(CHO)2 |
Quantity | 1 g |
Formula Weight | 150.95 |
Percent Purity | 97+% |
Chemical Name or Material | 2-Bromomalonaldehyde |
Safety and Handling
Product Identifier
- 2-Bromomalonaldehyde
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- H318-Causes serious eye damage.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'