missing translation for 'onlineSavingsMsg'
Learn More

2-(Bromomethyl)naphthalene, 96%, Thermo Scientific Chemicals

Product Code. 11400418
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water (reacts), and chloroform.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 939-26-4
Molecular Formula C11H9Br
Molecular Weight (g/mol) 221.10
MDL Number MFCD00004123
InChI Key RUHJZSZTSCSTCC-UHFFFAOYSA-N
Synonym 2-bromomethyl naphthalene, naphthalene, 2-bromomethyl, 2-naphthylmethyl bromide, 2-naphthyl bromomethane, 2-bromomethyl napthalene, .beta.-bromomethyl naphthalene, 2-brommethyl naphthalen, zlchem 566, 2-menaphthyl bromide
PubChem CID 70320
IUPAC Name 2-(bromomethyl)naphthalene
SMILES BrCC1=CC=C2C=CC=CC2=C1

Specifications

Melting Point 51°C to 55°C
Boiling Point 213°C (100 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
UN Number UN3261
Beilstein 636546
Solubility Information Soluble in water (reacts),and chloroform.
Formula Weight 221.1
Percent Purity 96%
Chemical Name or Material 2-(Bromomethyl)naphthalene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'