missing translation for 'onlineSavingsMsg'
Learn More
2-Bromonaphthalene, 98+%, Thermo Scientific Chemicals Product Code.: 11478166

2-Bromonaphthalene, 98+%, Thermo Scientific Chemicals

Product Code. 11478166
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11478166

Brand: Thermo Scientific Alfa Aesar A15300.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2-Bromonaphthalene is used as a raw material in the preparation of biaryls through Suzuki cross coupling reaction. Further, it plays a vital role in the preparation of dyes. It is also used to study potential tumorigenicity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromonaphthalene is used as a raw material in the preparation of biaryls through Suzuki cross coupling reaction. Further, it plays a vital role in the preparation of dyes. It is also used to study potential tumorigenicity.

Solubility
Slightly soluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 580-13-2
Molecular Formula C10H7Br
Molecular Weight (g/mol) 207.07
MDL Number MFCD00004051
InChI Key APSMUYYLXZULMS-UHFFFAOYSA-N
PubChem CID 11372
IUPAC Name 2-bromonaphthalene
SMILES BrC1=CC=C2C=CC=CC2=C1

Specifications

Melting Point 53°C to 57°C
Density 1.605
Boiling Point 281°C to 282°C
Flash Point 112°C (233°F)
Odor Odorless
Quantity 5 g
Beilstein 1858110
Merck Index 14,1426
Synonym naphthalene, 2-bromo, 2-naphthyl bromide, 2-bromo-naphthalene, beta-bromonaphthalene, beta-naphthyl bromide, .beta.-bromonaphthalene, unii-syu33i753n, naphthalene,2-bromo, .beta.-naphthyl bromide, pbn-2
Solubility Information Slightly soluble in water.
Formula Weight 207.08
Percent Purity ≥98%
Chemical Name or Material 2-Bromonaphthalene
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberK28X003 Certificate TypeCertificate of Analysis Date14/01/2025 Catalog NumberA15300.06,A15300.14,A15300.22
Lot Number10170183 Certificate TypeCertificate of Analysis Date13/07/2022 Catalog NumberA15300.06,A15300.14,A15300.22
Lot Number10162981 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15300.06,A15300.14,A15300.22
Lot NumberK28X003 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15300.06,A15300.14,A15300.22
Lot NumberD5223A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15300.06,A15300.14,A15300.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Bromonaphthalene, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.