Learn More
2-Bromopropane, 99%, Thermo Scientific Chemicals
£53.20 - £1116.00
Chemical Identifiers
CAS | 75-26-3 |
---|---|
Molecular Formula | C3H7Br |
Molecular Weight (g/mol) | 122.99 |
InChI Key | NAMYKGVDVNBCFQ-UHFFFAOYSA-N |
Synonym | isopropyl bromide, propane, 2-bromo, isopropylbromide, 2-bromo-propane, sec-propyl bromide, unii-r651xov97z, ccris 7919, hsdb 623, i-propylbromide, 2-brompropan |
PubChem CID | 6358 |
IUPAC Name | 2-bromopropane |
SMILES | CC(C)Br |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10070700
|
Thermo Scientific Acros
107322500 |
250 mL | Glass bottle |
£53.20
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10060700
|
Thermo Scientific Acros
107320010 |
1 L | Glass bottle |
£155.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11914491
|
Thermo Scientific Acros
107320025 |
2.5 L | Glass bottle |
£321.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15179153
|
Thermo Scientific Acros
107320100 |
10 L | Plastic Drum |
£1116.00
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
75-26-3 | |
122.99 | |
isopropyl bromide, propane, 2-bromo, isopropylbromide, 2-bromo-propane, sec-propyl bromide, unii-r651xov97z, ccris 7919, hsdb 623, i-propylbromide, 2-brompropan | |
2-bromopropane |
C3H7Br | |
NAMYKGVDVNBCFQ-UHFFFAOYSA-N | |
6358 | |
CC(C)Br |
Specifications
75-26-3 | |
100.0 | |
Brown to Colorless | |
59.0°C | |
Authentic | |
C3H7Br | |
1.4250 to 1.4270 | |
09,69 | |
1.31 | |
Solubility in water: 0.3g/100mL. Other solubilities: miscible with alcohol,benzene,chloroform,ether,and acetone | |
CC(C)Br | |
122.99 | |
122.99 | |
Liquid |
98.5 | |
-89.0°C | |
1.3100g/mL | |
1°C | |
98.5% min. (GC) | |
(CH3)2CHBr | |
01,108 | |
15,5256 | |
isopropyl bromide, propane, 2-bromo, isopropylbromide, 2-bromo-propane, sec-propyl bromide, unii-r651xov97z, ccris 7919, hsdb 623, i-propylbromide, 2-brompropan | |
NAMYKGVDVNBCFQ-UHFFFAOYSA-N | |
2-bromopropane | |
6358 | |
99% | |
2-Bromopropane |
Safety and Handling
GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
May damage fertility.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Do not breathe dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 200-855-1
RUO – Research Use Only