missing translation for 'onlineSavingsMsg'
Learn More

2-Bromopropene, 99%, stab., Thermo Scientific Chemicals

Product Code. 11329065
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2-Bromopropene is used as a solvent and as an intermediate for organic synthesis. It is also used as a fumigant. Further, it reacts with nonanal to prepare 2-methyl-undec-1-en-3-ol using chromium(II) chloride as a reagent. It is utilized as a refrigerant used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromopropene is used as a solvent and as an intermediate for organic synthesis. It is also used as a fumigant. Further, it reacts with nonanal to prepare 2-methyl-undec-1-en-3-ol using chromium(II) chloride as a reagent. It is utilized as a refrigerant used in organic synthesis.

Solubility
Immiscible with water.

Notes
Light sensitive. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 557-93-7
Molecular Formula C3H5Br
Molecular Weight (g/mol) 120.977
MDL Number MFCD00000140
InChI Key PHMRPWPDDRGGGF-UHFFFAOYSA-N
Synonym 2-bromopropene, isopropenyl bromide, 1-propene, 2-bromo, 2-bromo-1-propene, 2-bromopropylene, propene, 2-bromo, isopropylene bromide, alpha-methylvinyl bromide, .alpha.-methylvinyl bromide, 2-bromo-propene
PubChem CID 11202
IUPAC Name 2-bromoprop-1-ene
SMILES CC(=C)Br

Specifications

Melting Point -87°C
Density 1.362
Boiling Point 47°C to 49°C
Flash Point 4°C (39°F)
Odor Pungent
Refractive Index 1.443
Quantity 25 g
UN Number UN1993
Beilstein 1731926
Sensitivity Light sensitive
Solubility Information Immiscible with water.
Formula Weight 120.98
Percent Purity 99%
Chemical Name or Material 2-Bromopropene, stabilized
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'