missing translation for 'onlineSavingsMsg'
Learn More

2-Bromopropionyl chloride, 98%, Thermo Scientific Chemicals

Product Code. 10626372
10 mL, Glass bottle
Click to view available options
Quantity:
10 mL
50 mL
250 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7148-74-5
Molecular Formula C3H4BrClO
Molecular Weight (g/mol) 171.42
MDL Number MFCD00000711
InChI Key OZGMODDEIHYPRY-UHFFFAOYNA-N
Synonym 2-bromopropionyl chloride, propanoyl chloride, 2-bromo, 2-bromopropionylchloride, 2-bromo-propionyl chloride, 2-brominepropionylchloride, alpha-bromopropionyl chloride, 2-bromopropionic acid chloride, sgqdjlpqbsmuh@, 2-bromopropanoylchloride, acmc-1cjhb
PubChem CID 97980
IUPAC Name 2-bromopropanoyl chloride
SMILES CC(Br)C(Cl)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Density 1.7000g/mL
Boiling Point 131°C to 133°C
Flash Point 51°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ex Cl) (Argentometry)
Packaging Glass bottle
Linear Formula CH3CH(Br)COCl
Refractive Index 1.4770 to 1.481
Quantity 10 mL
Beilstein 02, 256
Specific Gravity 1.7
Solubility Information Solubility in water: reacts
Formula Weight 171.42
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Bromopropionyl chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Bromopropionyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Bromopropionyl chloride, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.