Learn More
2-Bromopyridine-3-carboxylic acid, 97%, Thermo Scientific™
£27.32 - £96.60
Chemical Identifiers
CAS | 35905-85-2 |
---|---|
Molecular Formula | C6H4BrNO2 |
Molecular Weight (g/mol) | 202.01 |
MDL Number | MFCD01646068 |
InChI Key | MMWNKXIFVYQOTK-UHFFFAOYSA-N |
Synonym | 2-bromonicotinic acid, 2-bromo-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 2-bromo, 2-bromo-nicotinic acid, 2-bromo-3-carboxypyridine, 3-bromopyridine-3-carboxylic acid, bromo-nicotinic acid, pubchem5071, acmc-1aj4p, ksc223s8f |
PubChem CID | 233262 |
IUPAC Name | 2-bromopyridine-3-carboxylic acid |
SMILES | C1=CC(=C(N=C1)Br)C(=O)O |
Description
Chemical Identifiers
35905-85-2 | |
202.01 | |
MMWNKXIFVYQOTK-UHFFFAOYSA-N | |
233262 | |
C1=CC(=C(N=C1)Br)C(=O)O |
C6H4BrNO2 | |
MFCD01646068 | |
2-bromonicotinic acid, 2-bromo-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 2-bromo, 2-bromo-nicotinic acid, 2-bromo-3-carboxypyridine, 3-bromopyridine-3-carboxylic acid, bromo-nicotinic acid, pubchem5071, acmc-1aj4p, ksc223s8f | |
2-bromopyridine-3-carboxylic acid |
Specifications
35905-85-2 | |
100.0 | |
96% min. (HPLC) | |
C6H4BrNO2 | |
2-bromonicotinic acid, 2-bromo-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 2-bromo, 2-bromo-nicotinic acid, 2-bromo-3-carboxypyridine, 3-bromopyridine-3-carboxylic acid, bromo-nicotinic acid, pubchem5071, acmc-1aj4p, ksc223s8f | |
C1=CC(=C(N=C1)Br)C(=O)O | |
202.01 | |
202.01 | |
2-Bromopyridine-3-carboxylic acid |
96.0 | |
200.0°C to 203.0°C | |
Glass Bottle | |
MFCD01646068 | |
MMWNKXIFVYQOTK-UHFFFAOYSA-N | |
2-bromopyridine-3-carboxylic acid | |
233262 | |
97% |
Safety and Handling
GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
- 2-Bromopyridine-3-carboxylic acid
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only