Learn More
2-Butanone, +99%, Extra Pure, Thermo Scientific Chemicals
£27.40 - £294.00
Chemical Identifiers
CAS | 78-93-3 |
---|---|
Molecular Formula | C4H8O |
Molecular Weight (g/mol) | 72.11 |
MDL Number | MFCD00011648 |
InChI Key | ZWEHNKRNPOVVGH-UHFFFAOYSA-N |
Synonym | 2-butanone, methyl ethyl ketone, butanone, ethyl methyl ketone, methylethyl ketone, meetco, methyl acetone, methylethylketone, 3-butanone, ethylmethylketon |
PubChem CID | 6569 |
ChEBI | CHEBI:28398 |
IUPAC Name | butan-2-one |
SMILES | CCC(C)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10502551
|
Thermo Scientific Acros
149670250 |
25 mL | Glass Bottle |
£27.40
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10306510
|
Thermo Scientific Acros
149670010 |
1 L | Glass Bottle |
£28.10
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10367320
|
Thermo Scientific Acros
149670025 |
2.5 L | Glass Bottle |
£52.80
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10227333
|
Thermo Scientific Acros
149670251 |
25 L | Metal Drum |
£294.00
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identificadores de productos químicos
78-93-3 | |
72.11 | |
ZWEHNKRNPOVVGH-UHFFFAOYSA-N | |
6569 | |
butan-2-one |
C4H8O | |
MFCD00011648 | |
2-butanone, methyl ethyl ketone, butanone, ethyl methyl ketone, methylethyl ketone, meetco, methyl acetone, methylethylketone, 3-butanone, ethylmethylketon | |
CHEBI:28398 | |
CCC(C)=O |
Especificaciones
-87.0°C | |
99.0 | |
100.0 | |
80.0°C | |
Authentic | |
1.3780 to 1.3800 | |
0.005% max. | |
C4H8O | |
MFCD00011648 | |
0.42 mPa.s (15°C) | |
0.2% max. | |
15, 6143 | |
Solubility in water: 290g/L (20°C). Other solubilities: soluble in benzene, alcohol and ether, miscible with oils | |
CCC(C)=O | |
72.11 | |
CHEBI:28398 | |
≥99% | |
2-Butanone |
78-93-3 | |
Colorless | |
0.8060g/mL | |
−7°C | |
Liquid | |
99+% | |
0.806 | |
C2H5COCH3 | |
Vapor Pressure: 105mbar at 20°C | |
01, IV, 3243 | |
01,678 | |
2-butanone, methyl ethyl ketone, butanone, ethyl methyl ketone, methylethyl ketone, meetco, methyl acetone, methylethylketone, 3-butanone, ethylmethylketon | |
ZWEHNKRNPOVVGH-UHFFFAOYSA-N | |
butan-2-one | |
6569 | |
72.11 | |
Extra Pure |
Seguridad y manipulación
GHS H Statement:
Causes serious eye irritation.
May cause drowsiness or dizziness.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
- 2-Butanone
Palabra de advertencia
- Danger
Clasificación
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Indicaciones de peligro
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Consejos de prudencia
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only