missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Butanone, Honeywell
Click to view available options
Quantity:
1 L
2.5 L
30 kg
Unit Size:
1 L
2.5 L
30kg
Specifications
Specifications
| Density | 0.805g/mL at 25°C |
| Quantity | 1 L |
| Explosion Limit | 0.101 |
| Flash Point | −3°C (26.6°F) |
| Ignition Point | 960°F |
| Linear Formula | C2H5COCH3 |
| Vapor Pressure | 71 mmHg (20°C) |
| UN Number | UN1193 |
| Beilstein | 741880 |
| Formula Weight | 72.11g/mol |
| Show More |
Safety and Handling
Product Identifier
- 2-Butanone
- Danger
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering