missing translation for 'onlineSavingsMsg'
Learn More
2-Butanone, for HPLC, Thermo Scientific Chemicals Product Code.: 10205613

2-Butanone, for HPLC, Thermo Scientific Chemicals

Product Code. 10205613
1 L, Glass Bottle
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 10205613

Brand: Thermo Scientific Acros 389570010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For HPLC
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-93-3
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00011648
InChI Key ZWEHNKRNPOVVGH-UHFFFAOYSA-N
Synonym 2-butanone, methyl ethyl ketone, butanone, ethyl methyl ketone, methylethyl ketone, meetco, methyl acetone, methylethylketone, 3-butanone, ethylmethylketonShow MoreShow Less
PubChem CID 6569
ChEBI CHEBI:28398
IUPAC Name butan-2-one
SMILES CCC(C)=O

Specifications

Melting Point -87.0°C
Absorbance 0.009 max. (1 cm cell vs water) at 350nm, 0.009 max. (1 cm cell vs water) at 450nm, 0.097 max. (1 cm cell vs water) at 340nm, 0.699 max. (1 cm cell vs water) at 330nm, 1 max. (1 cm cell vs water) at 329nm
Acidity 0.0003 meq/g max.
Density 0.8060g/mL
Boiling Point 80.0°C
Quantity 1 L
Flash Point −7°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3780 to 1.3800
Residue after Evaporation 0.001% max.
Specific Gravity 0.806
Linear Formula C2H5COCH3
Viscosity 0.42 mPa.s (15°C)
Water 0.1% max.
Merck Index 15, 6143
Solubility Information Solubility in water: 290g/L (20°C). Other solubilities: soluble in benzene, alcohol and ether, miscible with oils
Formula Weight 72.11
Percent Purity 99.7%
Grade HPLC
Chemical Name or Material 2-Butanone
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0462161 Certificate TypeCertificate of Analysis Date29/04/2024 Catalog Number389570010,389570025
Lot NumberA0459091 Certificate TypeCertificate of Analysis Date05/01/2024 Catalog Number389570010,389570025
Lot NumberA0443001 Certificate TypeCertificate of Analysis Date15/12/2023 Catalog Number389570010,389570025
Lot NumberA0394988 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number389570010,389570025
Lot NumberA0404233 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number389570010,389570025
compliance-icons compliance-icons
Product Identifier
  • 2-Butanone
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Methyl ethyl ketone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Butanone, for HPLC, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.