missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Butanone, Puriss., ≥99% (GC), Honeywell
Click to view available options
Quantity:
2.5 L
Packaging:
Glass Bottle
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Specifications
Melting Point | -87°C (literature) |
Density | 0.805 g/mL at 25 °C(lit.) |
Boiling Point | 79°C to 81°C |
Quantity | 2.5 L |
Flash Point | −4°C |
Packaging | Glass Bottle |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'