missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Butanone, puriss. p.a. ACS Reagent, ≥99.5% (GC), Honeywell Riedel-de Haën™
missing translation for 'orderingAttributeHoverText'
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Descrizione
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifica
Melting Point | -87°C |
Appearance | Colorless |
Density | 0.805g/mL at 25°C |
Boiling Point | 79°C to 81°C |
Quantity | 1 L |
Flash Point | −3°C (26.6°F) |
Packaging | Glass Bottle |
Refractive Index | n20/D 1.379(lit.) |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'