missing translation for 'onlineSavingsMsg'
Learn More

2-Butanone, ReagentPlus™, ≥99%, Honeywell™

Product Code. 15601010
Click to view available options
Quantity:
500 mL
1 L
2.5 L
25 L
4 x 2.5 L
Packaging:
Glass Bottle
Stainless Steel Can
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Specifications

Melting Point -87°C
Appearance Colorless
Density 0.805g/mL at 25°C
Boiling Point 79°C to 81°C
Quantity 25 L
Flash Point −3°C (26.6°F)
Packaging Stainless Steel Can
Refractive Index n20/D 1.379(lit.)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.