missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Butanone, ReagentPlus™, ≥99%, Honeywell™
Click to view available options
Quantity:
500 mL
1 L
2.5 L
25 L
4 x 2.5 L
Packaging:
Glass Bottle
Stainless Steel Can
Description
- ≥95% for general laboratory use.
Specifications
Specifications
Melting Point | -87°C |
Appearance | Colorless |
Density | 0.805g/mL at 25°C |
Boiling Point | 79°C to 81°C |
Quantity | 1 L |
Flash Point | −4°C (26.6°F) |
Packaging | Glass Bottle |
Refractive Index | n20/D 1.379(lit.) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction