missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Butenylmagnesium chloride, 0.5M in MeTHF, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
500 mL
Description
2-Butenylmagnesium chloride is used as a Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Butenylmagnesium chloride is used as a Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF).
Solubility
It reacts violently with water.
Notes
Air & moisture sensitive. Incompatible with oxidizing agents and bases.
2-Butenylmagnesium chloride is used as a Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF).
Solubility
It reacts violently with water.
Notes
Air & moisture sensitive. Incompatible with oxidizing agents and bases.
Specifications
Specifications
Linear Formula | CH3CH=CHCH2MgCl |
Quantity | 500 mL |
UN Number | UN3399 |
InChI Key | RTJGBPYEBKZQGE-CZEFNJPISA-M |
SMILES | C\C=C\C[Mg]Cl |
IUPAC Name | (2E)-3-(4-chlorophenyl)-2-[(furan-2-yl)formamido]prop-2-enamide |
Molecular Weight (g/mol) | 114.86 |
Formula Weight | 114.86 |
Concentration or Composition (by Analyte or Components) | 0.5M in MeTHF |
Chemical Name or Material | 2-Butenylmagnesium chloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'