missing translation for 'onlineSavingsMsg'
Learn More

2-Butoxyethanol, 99%, extra pure, Thermo Scientific Chemicals

£34.67 - £381.90

Chemical Identifiers

CAS 111-76-2
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.18
MDL Number MFCD00002884
InChI Key POAOYUHQDCAZBD-UHFFFAOYSA-N
Synonym butoxyethanol, butyl glycol, butyl cellosolve, ethylene glycol monobutyl ether, n-butoxyethanol, ethanol, 2-butoxy, ethylene glycol butyl ether, butyl oxitol, glycol butyl ether, glycol ether eb
PubChem CID 8133
ChEBI CHEBI:63921
SMILES CCCCOCCO
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10532171
View Documents Promotion Details
Thermo Scientific Acros
154330010
1 L Glass Bottle
£34.67
1 L
Please to purchase this item. Need a web account? Register with us today!
10448690
View Documents
Thermo Scientific Acros
154330025
2.5 L Glass Bottle
£65.73
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10217503
View Documents
Thermo Scientific Acros
154330100
10 L Plastic drum
£190.95
10 L
Please to purchase this item. Need a web account? Register with us today!
10092103
View Documents
Thermo Scientific Acros
154330250
25 L Metal drum
£381.90
25 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

111-76-2
118.18
POAOYUHQDCAZBD-UHFFFAOYSA-N
8133
CCCCOCCO
C6H14O2
MFCD00002884
butoxyethanol, butyl glycol, butyl cellosolve, ethylene glycol monobutyl ether, n-butoxyethanol, ethanol, 2-butoxy, ethylene glycol butyl ether, butyl oxitol, glycol butyl ether, glycol ether eb
CHEBI:63921

Specifications

111-76-2
100.0
0.9010g/mL
62°C
98.5% min. (GC)
CH3(CH2)3OCH2CH2OH
MFCD00002884
0.02% max.
0.901
Solubility in water: soluble. Other solubilities: soluble in most organic solvents
CCCCOCCO
118.18
CHEBI:63921
118.18
Extra Pure
2-Butoxyethanol
98.5
-70.0°C
171.0°C
Authentic
C6H14O2
1.4180 to 1.4200
01, II, 519
15, 1560
butoxyethanol, butyl glycol, butyl cellosolve, ethylene glycol monobutyl ether, n-butoxyethanol, ethanol, 2-butoxy, ethylene glycol butyl ether, butyl oxitol, glycol butyl ether, glycol ether eb
POAOYUHQDCAZBD-UHFFFAOYSA-N
2-butoxyethan-1-ol
8133
5.31 mPa.s (20°C)
99%
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed: Call a POISON CENTER or doctor/physician.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously wit

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 203-905-

Product Identifier
  • 2-Butoxyethanol

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.