missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Butoxyethyl acetate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 L
2.5 L
10 L
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Melting Point | -64.0°C |
Density | 0.9385g/mL |
Boiling Point | 193.5°C |
Flash Point | 76°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (GC) |
Packaging | Glass bottle |
Linear Formula | CH3CO2CH2CH2O(CH2)3CH3 |
Show More |
Safety and Handling
Product Identifier
- 2-Butoxyethyl acetate
- Warning
- Acute toxicity Category 4
- H332-Harmful if inhaled.
- H312-Harmful in contact with skin.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'